Learn More
Cayman Chemical 3 4-MDPAhydrochlorIde 10mg
SDP
Catalog No. 502954306
Encompass_Preferred
A lesser known psychoactive substituted amphetamine of the phenethylamine class that has been used as an internal standard for qualitative and quantitative analyses of hallucinogenic designer drugs in biological samples; intended for forensic and research purposes
Catalog No. 50-295-4306 Supplier Cayman Chemical Supplier No. 1397210 MG
A lesser known psychoactive substituted amphetamine of the phenethylamine class that has been used as an internal standard for qualitative and quantitative analyses of hallucinogenic designer drugs in biological samples; intended for forensic and research purposes