Learn More
Sigma Aldrich 3-(4-Methoxy-benzyl)-piperidine
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Tandem mass spectrometry data independently generated by scripps center for metabolomics is available to view or download in pdf
- M47353.pdf tested metabolites are featured on scripps center for metabolomics metlin metabolite database
- To learn more, visit sigma.com/metlin
Specifications
Specifications
| Molecular Formula | C13H19NO |
| Linear Formula | C13H19NO |
| MDL Number | MFCD05863592 |
| Quantity | 25 g |
| Molecular Weight (g/mol) | 205.3 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.