Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4-Methoxyphenyl)-1-propylboronic acid pinacol ester, 97%

Catalog No. AAH2855706
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2855706 5 g
AAH28557MD 250 mg
AAH2855703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2855706 Supplier Thermo Scientific Chemicals Supplier No. H2855706
Add to Cart

CAS: 1073371-72-8 | C16H25BO3 | 276.183 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073371-72-8
Molecular Formula C16H25BO3
Molecular Weight (g/mol) 276.183
MDL Number MFCD09953503
InChI Key ZJNJYHQBUIHZLN-UHFFFAOYSA-N
Synonym 3-4-methoxyphenyl-1-propylboronic acid pinacol ester, 2-3-4-methoxyphenyl propyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-4-methoxyphenyl propylboronic acid pinacol ester, 3-4-methoxyphenyl propylboronic acid,pinacol ester, 3-4-methoxyphenyl propylboronic acid, pinacol ester
PubChem CID 46739066
IUPAC Name 2-[3-(4-methoxyphenyl)propyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)CCCC2=CC=C(C=C2)OC

Specifications

Quantity 5 g
Formula Weight 276.19
Percent Purity 97%
Chemical Name or Material 3-(4-Methoxyphenyl)-1-propylboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.