Learn More
Medchemexpress LLC 3-(4-methoxyphenyl)-5-[(5-nitro-2-thienyl)methylene]-2-thioxo-4-thiazolidinone | 346640-08-2 | 98.0% | 378.45 | C15H10N2O4S3 | 25 MG
SDP
Catalog No. 5000214665
Encompass_Preferred
CCF642 is a small-molecule protein disulfide isomerase (PDI) inhibitor reported to have an IC50 of 2.9 μM and to induce endoplasmic reticulum stress and apoptosis in multiple myeloma cells. The compound is supplied for research use with documented solubility and storage guidance for experimental workflows.
  • Potent PDI inhibition with reported IC50 of 2.9 μM.
  • Induces ER stress and apoptosis in multiple myeloma models.
  • High solubility in DMSO for in vitro stock preparation.
  • Stable as a powder under recommended cold storage conditions.
  • Documented in vitro and in vivo formulation guidance for experimental use.
Catalog No. 50-002-14665 Supplier Medchemexpress LLC Supplier No. HY10043025MG
CCF642 is a small-molecule protein disulfide isomerase (PDI) inhibitor reported to have an IC50 of 2.9 μM and to induce endoplasmic reticulum stress and apoptosis in multiple myeloma cells. The compound is supplied for research use with documented solubility and storage guidance for experimental workflows.
  • Potent PDI inhibition with reported IC50 of 2.9 μM.
  • Induces ER stress and apoptosis in multiple myeloma models.
  • High solubility in DMSO for in vitro stock preparation.
  • Stable as a powder under recommended cold storage conditions.
  • Documented in vitro and in vivo formulation guidance for experimental use.