Learn More
Medchemexpress LLC 3-(4-methoxyphenyl)-5-[(5-nitro-2-thienyl)methylene]-2-thioxo-4-thiazolidinone | 346640-08-2 | MFCD01899742 | 98.0% | 378.45 | C15H10N2O4S3 | 2 MG
SDP
Catalog No. 5000214875
Encompass_Preferred
CCF642 is a small-molecule protein disulfide isomerase (PDI) inhibitor used in biochemical and cell-based research. It has a reported IC50 of ≈2.9 μM and can induce acute endoplasmic reticulum stress. The compound is supplied in solid and solution formats for laboratory experiments and is intended for research use only.
  • Potent PDI inhibitor with reported IC50 ≈2.9 μM.
  • Induces acute endoplasmic reticulum stress in cell models.
  • Available as solid powder and DMSO solution formats.
  • High purity, typically around 98.0%.
  • Molecular weight 378.45 g/mol.
  • Intended for research use only; not for human or animal therapeutic use.
Catalog No. 50-002-14875 Supplier Medchemexpress LLC Supplier No. HY1004302MG
CCF642 is a small-molecule protein disulfide isomerase (PDI) inhibitor used in biochemical and cell-based research. It has a reported IC50 of ≈2.9 μM and can induce acute endoplasmic reticulum stress. The compound is supplied in solid and solution formats for laboratory experiments and is intended for research use only.
  • Potent PDI inhibitor with reported IC50 ≈2.9 μM.
  • Induces acute endoplasmic reticulum stress in cell models.
  • Available as solid powder and DMSO solution formats.
  • High purity, typically around 98.0%.
  • Molecular weight 378.45 g/mol.
  • Intended for research use only; not for human or animal therapeutic use.