Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(4-Methoxyphenyl)propionic acid

Catalog No. M23527100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23527100G 100 g
M2352710G 10 g
M2352725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M23527100G Supplier Sigma Aldrich Supplier No. M23527100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-(4-Methoxyphenyl)propionic acid

Specifications

CAS 1929-29-9
Melting Point 98°C to 100°C (lit.)
Molecular Formula C10H12O3
Linear Formula CH3OC6H4CH2CH2CO2H
MDL Number MFCD00002777
Quantity 100 g
Synonym 4-Methoxyhydrocinnamic acid
Molecular Weight (g/mol) 180.2
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.