Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-[4-methyl-2-(2-oxo-1,2-dihydro-indol-3-ylidenemethyl)-1H-pyrrol-3-yl]-propionic acid | 215543-92-3 | MFCD08235144 | 99.9% | 296.32 g·mol⁻¹ | C17H16N2O3 | 1 MG
SDP

Catalog No. 5000242521
Encompass_Preferred

SU 5402 is a small-molecule receptor tyrosine kinase inhibitor used in research to probe FGFR, VEGFR2, and PDGFRβ signaling. Supplied as a purified solid for in vitro applications, it exhibits low-nanomolar potency against VEGFR2 and FGFR1 and sub-micromolar activity against PDGFRβ.

  • Inhibits VEGFR2, FGFR1, and PDGFRβ (IC50: 20 nM, 30 nM, 510 nM).
  • Supplied as a high-purity solid suitable for biochemical and cell-based assays.
  • Molecular weight 296.32 g·mol⁻¹; formula C17H16N2O3.
  • Recommended storage: powder frozen (-20°C) and solutions at -80°C for long-term stability.
  • Commonly used to study angiogenesis and receptor tyrosine kinase signaling pathways.

Catalog No. 50-002-42521 Supplier Medchemexpress LLC Supplier No. HY104071MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SU 5402 is a small-molecule receptor tyrosine kinase inhibitor used in research to probe FGFR, VEGFR2, and PDGFRβ signaling. Supplied as a purified solid for in vitro applications, it exhibits low-nanomolar potency against VEGFR2 and FGFR1 and sub-micromolar activity against PDGFRβ.

  • Inhibits VEGFR2, FGFR1, and PDGFRβ (IC50: 20 nM, 30 nM, 510 nM).
  • Supplied as a high-purity solid suitable for biochemical and cell-based assays.
  • Molecular weight 296.32 g·mol⁻¹; formula C17H16N2O3.
  • Recommended storage: powder frozen (-20°C) and solutions at -80°C for long-term stability.
  • Commonly used to study angiogenesis and receptor tyrosine kinase signaling pathways.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.