Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Methylenedioxy-N-ethylaniline 98.0+%, TCI America™
SDP

Catalog No. M094825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M094825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M094825G Supplier TCI America Supplier No. M094825G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 32953-14-3
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00009743
InChI Key FPKGTVXPIULTIP-UHFFFAOYSA-N
Synonym n-ethyl-3,4-methylenedioxy aniline, 3,4-methylenedioxy-n-ethylaniline, n-ethylbenzo d 1,3 dioxol-5-amine, 1,3-benzodioxol-5-amine, n-ethyl, n-ethyl-3,4-methylenedioxyaniline, n-ethyl-2h-1,3-benzodioxol-5-amine, n-ethylbenzo-1,3-dioxol-5-amine, acmc-1admb, 5-ethylamino-1,3-benzodioxole, fpkgtvxpiultip-uhfffaoysa
PubChem CID 118365
IUPAC Name N-ethyl-1,3-benzodioxol-5-amine
SMILES CCNC1=CC2=C(C=C1)OCO2

Specifications

Boiling Point 122°C
Quantity 25 g
Formula Weight 165.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3,4-Methylenedioxy-N-ethylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.