Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-(Methylenedioxy)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. M203525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2035-25G 25 g
M20355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2035-25G Supplier TCI America Supplier No. M203525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 94839-07-3
Melting Point 229°C
Color White-Yellow
Molecular Formula C7H7BO4
Quantity 25 g
MDL Number MFCD01009695
Synonym 3,4-methylenedioxyphenylboronic acid, benzo d 1,3 dioxol-5-ylboronic acid, 3,4-methylenedioxy phenylboronic acid, 3,4-methylenedioxyphenyl boronic acid, 3,4-methylenedioxy benzeneboronic acid
InChI Key CMHPUBKZZPSUIQ-UHFFFAOYSA-N
SMILES B(C1=CC2=C(C=C1)OCO2)(O)O
IUPAC Name 1,3-benzodioxol-5-ylboronic acid
Molecular Weight (g/mol) 165.939
PubChem CID 2734371
Formula Weight 165.94
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.