Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,4-methylenedioxy PV9 hydrochloride | 24646-40-0 | 99.9% | 353.88 | C19H28ClNO3 | 1 ML
SDP

Catalog No. 5000227499
Encompass_Preferred

3,4-Methylenedioxy PV9 hydrochloride (CAS 24646-40-0) is a research-grade cathinone derivative supplied as a ready-to-use 10 mM solution in DMSO and as solid material. It is intended for laboratory research and analytical applications and should be handled according to the safety data sheet.

  • Ready-to-use 10 mM solution in DMSO, 1 mL volume.
  • High purity: 99.9%.
  • Available as solid in multiple sizes (5 mg-500 mg).
  • Molecular weight 353.88 and formula C19H28ClNO3.
  • Storage: solid at 4°C; in solvent -80°C up to 6 months or -20°C up to 1 month.
  • For research and analytical use only; not for human or veterinary use.

Catalog No. 50-002-27499 Supplier Medchemexpress LLC Supplier No. HY11882910MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3,4-Methylenedioxy PV9 hydrochloride (CAS 24646-40-0) is a research-grade cathinone derivative supplied as a ready-to-use 10 mM solution in DMSO and as solid material. It is intended for laboratory research and analytical applications and should be handled according to the safety data sheet.

  • Ready-to-use 10 mM solution in DMSO, 1 mL volume.
  • High purity: 99.9%.
  • Available as solid in multiple sizes (5 mg-500 mg).
  • Molecular weight 353.88 and formula C19H28ClNO3.
  • Storage: solid at 4°C; in solvent -80°C up to 6 months or -20°C up to 1 month.
  • For research and analytical use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.