Learn More
Medchemexpress LLC 3,4-methylenedioxy PV9 hydrochloride | 24646-40-0 | 99.9% | 353.88 | C19H28ClNO3 | 10 MG
SDP
Catalog No. 5000229006
Encompass_Preferred
3,4-Methylenedioxy PV9 hydrochloride is the hydrochloride salt of the PV9 derivative supplied as a high-purity analytical reference for laboratory research and assay development. It is provided as a solid and offered in small pack sizes suitable for analytical use.
  • High purity: 99.9%.
  • Solid crystalline form for ease of handling.
  • Available in small pack sizes including 10 mg for analytical workflows.
  • Storage: keep sealed and away from moisture; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).
Catalog No. 50-002-29006 Supplier Medchemexpress LLC Supplier No. HY11882910MG
3,4-Methylenedioxy PV9 hydrochloride is the hydrochloride salt of the PV9 derivative supplied as a high-purity analytical reference for laboratory research and assay development. It is provided as a solid and offered in small pack sizes suitable for analytical use.
  • High purity: 99.9%.
  • Solid crystalline form for ease of handling.
  • Available in small pack sizes including 10 mg for analytical workflows.
  • Storage: keep sealed and away from moisture; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).