Learn More
Medchemexpress LLC 3,4-methylenedioxy PV9 hydrochloride | 24646-40-0 | 99.9% | 353.88 | C19H28ClNO3 | 100 MG
SDP
Catalog No. 5000227385
Encompass_Preferred
3,4-Methylenedioxy PV9 hydrochloride is the hydrochloride salt of the PV9 drug derivative supplied for laboratory research use. The compound is reported to show in vitro cytotoxicity against multiple cell lines with LC50 values in the 12.8-67.5 μM range. Intended for research use only; not for human consumption.
  • Hydrochloride salt of a cathinone-type PV9 derivative.
  • Demonstrated in vitro cytotoxicity across several cell lines (LC50 12.8-67.5 μM).
  • High purity typical of analytical standards (>99%).
  • Crystalline solid, white to light brown appearance.
  • Storage: 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month).
  • Provided as a 100 mg research-pack quantity.
Catalog No. 50-002-27385 Supplier Medchemexpress LLC Supplier No. HY118829100MG
3,4-Methylenedioxy PV9 hydrochloride is the hydrochloride salt of the PV9 drug derivative supplied for laboratory research use. The compound is reported to show in vitro cytotoxicity against multiple cell lines with LC50 values in the 12.8-67.5 μM range. Intended for research use only; not for human consumption.
  • Hydrochloride salt of a cathinone-type PV9 derivative.
  • Demonstrated in vitro cytotoxicity across several cell lines (LC50 12.8-67.5 μM).
  • High purity typical of analytical standards (>99%).
  • Crystalline solid, white to light brown appearance.
  • Storage: 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month).
  • Provided as a 100 mg research-pack quantity.