Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,4-methylenedioxy PV9 hydrochloride | 24646-40-0 | 99.9% | 353.88 g/mol | C19H28ClNO3 | 25 MG
SDP

Catalog No. 5000307905
Encompass_Preferred

3,4-Methylenedioxy PV9 hydrochloride is a crystalline hydrochloride salt supplied as a white to light brown solid for laboratory research and analytical use only. It is provided in small pack sizes for assay and reference applications.

  • High purity: 99.9%.
  • Molecular weight: 353.88 g/mol.
  • Chemical formula: C19H28ClNO3.
  • CAS number: 24646-40-0.
  • Available in small pack sizes including 25 mg suitable for analytical work.

Catalog No. 50-003-07905 Supplier Medchemexpress LLC Supplier No. HY11882925MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3,4-Methylenedioxy PV9 hydrochloride is a crystalline hydrochloride salt supplied as a white to light brown solid for laboratory research and analytical use only. It is provided in small pack sizes for assay and reference applications.

  • High purity: 99.9%.
  • Molecular weight: 353.88 g/mol.
  • Chemical formula: C19H28ClNO3.
  • CAS number: 24646-40-0.
  • Available in small pack sizes including 25 mg suitable for analytical work.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.