Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,4-Methylenedioxy PV9 hydrochloride | 24646-40-0 | 99.9% | 353.88 g·mol⁻¹ | C19H28ClNO3 | 5 MG
SDP

Catalog No. 5000227380
Encompass_Preferred

3,4-Methylenedioxy PV9 hydrochloride is a hydrochloride salt of a PV9 derivative provided as a solid research reagent for in vitro studies and reference standard applications. It is characterized by a defined molecular formula and molecular weight, and is supplied with manufacturer guidance for storage and handling.

  • CAS number 24646-40-0.
  • Molecular weight 353.88 g·mol⁻¹.
  • Chemical formula C19H28ClNO3.
  • Form crystalline solid, hydrochloride salt.
  • Packaged quantity 5 MG.
  • Purity 99.9% (HPLC).
  • Storage sealed, away from moisture; in solvent: -80 °C up to 6 months, -20 °C up to 1 month.

Catalog No. 50-002-27380 Supplier Medchemexpress LLC Supplier No. HY1188295MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3,4-Methylenedioxy PV9 hydrochloride is a hydrochloride salt of a PV9 derivative provided as a solid research reagent for in vitro studies and reference standard applications. It is characterized by a defined molecular formula and molecular weight, and is supplied with manufacturer guidance for storage and handling.

  • CAS number 24646-40-0.
  • Molecular weight 353.88 g·mol⁻¹.
  • Chemical formula C19H28ClNO3.
  • Form crystalline solid, hydrochloride salt.
  • Packaged quantity 5 MG.
  • Purity 99.9% (HPLC).
  • Storage sealed, away from moisture; in solvent: -80 °C up to 6 months, -20 °C up to 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.