Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(4-Methylthiazol-2-yl)propan-1-amine

Catalog No. OTV0006041G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OTV0006041G 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OTV0006041G Supplier Sigma Aldrich Supplier No. OTV0006041G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,2-Dichlorobutane causes the alkylation of benzene using aluminium chloride or liquid hydrogen fluoride as catalyst.1,2-Dichlorobutane was used to evaluate temperature-composition phase diagrams of binary mixtures composed of a haloalkane dissolved in either 1-ethyl-3-methylimidazolium ethylsulfate or 1-ethyl-3-methylimidazolium ethylsulfonate.

Specifications

CAS 112086-66-5
Molecular Formula C7H12N2S
Linear Formula C7H12N2S
MDL Number MFCD09891572
Quantity 10 mL
Molecular Weight (g/mol) 156.25
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.