Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-[4-(Methylthio)phenyl]propionic acid, 96%

Catalog No. AAH3410103
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3410103 1 g
AAH3410109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3410103 Supplier Thermo Scientific Chemicals Supplier No. H3410103
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 138485-81-1 | C10H12O2S | 196.264 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138485-81-1
Molecular Formula C10H12O2S
Molecular Weight (g/mol) 196.264
MDL Number MFCD03425696
InChI Key ASTVFRXZLBGVRL-UHFFFAOYSA-N
PubChem CID 3801098
IUPAC Name 3-(4-methylsulfanylphenyl)propanoic acid
SMILES CSC1=CC=C(C=C1)CCC(=O)O

Specifications

Melting Point 97°C to 102°C
Quantity 1 g
Synonym 3-4-methylsulfanyl phenyl propanoic acid, 3-4-methylthio phenyl propanoic acid, 3-4-methylthiophenyl propionic acid, benzenepropanoicacid, 4-methylthio, benzenepropanoic acid, 4-methylthio, 3-4-methylthio phenyl propionic acid, 3-4-methylsulfanylphenyl propanoic acid, acmc-20ex3q, benzenepropanoicacid,4-methylthio
Formula Weight 196.26
Percent Purity 96%
Chemical Name or Material 3-[4-(Methylthio)phenyl]propionic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.