Learn More
Medchemexpress LLC 3-(4-morpholinopyrido[3',2':4,5]furo[3,2-d]pyrimidin-2-yl)phenol | 371935-74-9 | MFCD11983145 | 99.0% | 348.36 g/mol | C19H16N4O3 | 50 MG
SDP
Catalog No. 5000213142
Encompass_Preferred
A small-molecule research reagent that potently inhibits PI3K and mTOR signaling and is used in biochemical and cell-based studies. It also inhibits DNA-PK and has been reported to induce autophagy.
  • Potent PI3K and mTOR inhibitor with reported low-nanomolar IC50s (p110α 8 nM; mTORC1 20 nM).
  • Also inhibits DNA-PK (IC50 2 nM).
  • Used for biochemical and cell-based research; for research use only.
  • Stable as a powder at -20°C (up to 3 years) or 4°C (up to 2 years).
  • Recommended in-solution storage: -80°C (1 year) or -20°C (6 months).
  • Supplied in milligram-scale quantities suitable for analytical and cellular studies.
Catalog No. 50-002-13142 Supplier Medchemexpress LLC Supplier No. HY1011550MG
A small-molecule research reagent that potently inhibits PI3K and mTOR signaling and is used in biochemical and cell-based studies. It also inhibits DNA-PK and has been reported to induce autophagy.
  • Potent PI3K and mTOR inhibitor with reported low-nanomolar IC50s (p110α 8 nM; mTORC1 20 nM).
  • Also inhibits DNA-PK (IC50 2 nM).
  • Used for biochemical and cell-based research; for research use only.
  • Stable as a powder at -20°C (up to 3 years) or 4°C (up to 2 years).
  • Recommended in-solution storage: -80°C (1 year) or -20°C (6 months).
  • Supplied in milligram-scale quantities suitable for analytical and cellular studies.