Learn More
Medchemexpress LLC 3-(4-nitrophenyl)-1-phenyl-2-pyrazolin-5-one | 34320-83-7 | 99.7% | 281.27 g/mol | C15H11N3O3 | 5 MG
SDP
Catalog No. 5000240260
Encompass_Preferred
TCS PrP Inhibitor 13 is a small-molecule antiprion compound that inhibits accumulation of protease-resistant prion protein in cellular assays. It has been reported to show potent cellular activity and is used as a research reagent for studies of prion biology and for screening antiprion compounds.
  • Potent antiprion activity with reported cellular IC50 ~3 nM.
  • High chemical purity (99.7%).
  • Chemical formula C15H11N3O3 and molecular weight 281.27 g/mol.
  • Suitable for prion biology research and antiprion compound screening.
  • Provided in milligram quantities for laboratory studies.
Catalog No. 50-002-40260 Supplier Medchemexpress LLC Supplier No. HY1076625MG
TCS PrP Inhibitor 13 is a small-molecule antiprion compound that inhibits accumulation of protease-resistant prion protein in cellular assays. It has been reported to show potent cellular activity and is used as a research reagent for studies of prion biology and for screening antiprion compounds.
  • Potent antiprion activity with reported cellular IC50 ~3 nM.
  • High chemical purity (99.7%).
  • Chemical formula C15H11N3O3 and molecular weight 281.27 g/mol.
  • Suitable for prion biology research and antiprion compound screening.
  • Provided in milligram quantities for laboratory studies.