Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4-Nitrophenyl)-1H-pyrazole, 97%, Thermo Scientific Chemicals

Catalog No. p-7031304
Change view
Click to view available options
Quantity:
2 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3233204 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3233204 Supplier Thermo Scientific Chemicals Supplier No. H3233204
Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20583-31-7
Molecular Formula C9H7N3O2
Molecular Weight (g/mol) 189.17
MDL Number MFCD00665859
InChI Key IPIYADCDDIUVPS-UHFFFAOYSA-N
Synonym 3-4-nitrophenyl-1h-pyrazole, 3-4-nitrophenyl pyrazole, 5-4-nitrophenyl-1h-pyrazole, 3-4-nitro-phenyl-1h-pyrazole, 5-4-nitrophenyl pyrazole, 1h-pyrazole, 3-4-nitrophenyl, 5-4-nitro-phenyl-1h-pyrazole, 3-4-nitrophenyl-2h-pyrazole, nitrophenylpyrazole, p-nitrophenylpyrazole
PubChem CID 2737071
IUPAC Name 5-(4-nitrophenyl)-1H-pyrazole
SMILES [O-][N+](=O)C1=CC=C(C=C1)C1=CC=NN1

Specifications

Melting Point 194°C to 195°C
Quantity 2 g
Formula Weight 189.18
Percent Purity 97%
Chemical Name or Material 3-(4-Nitrophenyl)-1H-pyrazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.