Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4-Nitrophenyl)propanoic acid, 98%

Catalog No. AC458201000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458201000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC458201000 Supplier Thermo Scientific Chemicals Supplier No. 458201000

missing translation for 'validMainframeMsg'

CAS: 16642-79-8 | C9H9NO4 | 195.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16642-79-8
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.17
MDL Number MFCD00126834
InChI Key VZOPVJNBOQOLPN-UHFFFAOYSA-N
Synonym 3-4-nitrophenyl propanoic acid, 3-4-nitrophenyl propionic acid, benzenepropanoic acid, 4-nitro, 3-4-nitrophenyl-propionic acid, 4-nitrobenzenepropanoic acid, p-nitrohydrozimtsaure, pubchem13615, p-nitrohydrocinnamic acid
PubChem CID 85526
IUPAC Name 3-(4-nitrophenyl)propanoic acid
SMILES OC(=O)CCC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 166.0°C to 168.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 100 g
Formula Weight 195.18
Percent Purity 98%
Chemical Name or Material 3-(4-Nitrophenyl)propanoic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.