Learn More
Ace Glass, Inc. PTFE insert, 3/4" NPT, #15 compression fitting12.5 to 14mm OD tubing
SDP
Catalog No. 50977909
Encompass
Compression fitting PTFE inserts for 3/4NPT to #15 Ace-Thred. Fits 9 to 10.5mm O.D. tubing. For use with ACE PTFE reaction vessel heads.
Catalog No. 50-977-909 Supplier Ace Glass, Inc. Supplier No. 1287114
Compression fitting PTFE inserts for 3/4NPT to #15 Ace-Thred. Fits 9 to 10.5mm O.D. tubing. For use with ACE PTFE reaction vessel heads.