Learn More
Ace Glass, Inc. PTFE insert, 3/4" NPT, #18 compression fitting16 to 17mm OD tubing
SDP
Catalog No. 50983475
Encompass
Compression fitting PTFE inserts for 3/4NPT to #18 Ace-Thred. Fits 16 to 17mm O.D. tubing. For use with ACE PTFE reaction vessel heads.
Catalog No. 50-983-475 Supplier Ace Glass, Inc. Supplier No. 1287118
Compression fitting PTFE inserts for 3/4NPT to #18 Ace-Thred. Fits 16 to 17mm O.D. tubing. For use with ACE PTFE reaction vessel heads.