Learn More
Medchemexpress LLC 3,4-O-Isopropylidene-shikimic acid | 183075-03-8 | 214.22 | 1 MG
SDP
Catalog No. 5000415768
Encompass_Preferred
3,4-O-Isopropylidene-shikimic acid is a natural product isolated from the whole plants of *Hypericum wightianum*. It exhibits anti-inflammatory effects and antioxidant activities. It is intended for research use only and not for sale to patients.
  • Natural product.
  • Isolated from *Hypericum wightianum*.
  • Possesses anti-inflammatory effects.
  • Exhibits antioxidant activities.
Catalog No. 50-004-15768 Supplier Medchemexpress LLC Supplier No. HYN17821MG
3,4-O-Isopropylidene-shikimic acid is a natural product isolated from the whole plants of *Hypericum wightianum*. It exhibits anti-inflammatory effects and antioxidant activities. It is intended for research use only and not for sale to patients.
  • Natural product.
  • Isolated from *Hypericum wightianum*.
  • Possesses anti-inflammatory effects.
  • Exhibits antioxidant activities.