Learn More
Medchemexpress LLC 3,4-O-Isopropylidene-shikimic acid | 183075-03-8 | 214.22 | 5 MG
SDP
Catalog No. 5000414647
Encompass_Preferred
3,4-O-Isopropylidene-shikimic acid is a natural product that can be isolated from the whole plants of *Hypericum wightianum*. It exhibits anti-inflammatory and antioxidant activities.
  • Natural product
  • Isolated from *Hypericum wightianum*
  • Anti-inflammatory effect
  • Antioxidant activities
  • Appearance: Solid
  • Color: Off-white to light yellow
Catalog No. 50-004-14647 Supplier Medchemexpress LLC Supplier No. HYN17825MG
3,4-O-Isopropylidene-shikimic acid is a natural product that can be isolated from the whole plants of *Hypericum wightianum*. It exhibits anti-inflammatory and antioxidant activities.
  • Natural product
  • Isolated from *Hypericum wightianum*
  • Anti-inflammatory effect
  • Antioxidant activities
  • Appearance: Solid
  • Color: Off-white to light yellow