Learn More
Bellco Glass, Inc. 3/4 Port Plug Assembly
SDP
Catalog No. 501944717
Encompass_Preferred
Used in 3, 7 and 10 liter bioreactors for 3/4: accessories in head plate. Includes o-ring and hex nut. MADE IN THE USA
Catalog No. 50-194-4717 Supplier Bellco Glass, Inc. Supplier No. 196401034
Used in 3, 7 and 10 liter bioreactors for 3/4: accessories in head plate. Includes o-ring and hex nut. MADE IN THE USA