Learn More
Enzo Life Sciences 3-(4-Pyridyl)-1H-indole (5mg). CAS: 7272-84-6
SDP
Supplier:  Enzo Life Sciences ALX270429M005
Encompass_Preferred
A cell permeable, selective, and ATP-competitive inhibitor of Rho kinase (ROCK) activity (IC50=25µM). Does not inhibit the activation of Rho nor does it affect the in vitro activities of MLCK, PKCα, and SAPK2a/p38α. Shown to be less potent than Y-27632 (ALX-270-333). Purity: ≥98% (HPLC). Appearance: Off-white to beige solid. Solubility: Soluble in DMSO or dimethyl formamide. Long Term Storage: -20°C.
Catalog No. 50-200-8553