Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4-Pyridyl)-L-alanine dihydrochloride, 97%, Thermo Scientific™

Catalog No. AAH63928MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63928MD 250 mg
AAH6392803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH63928MD Supplier Thermo Scientific Chemicals Supplier No. H63928MD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 178933-04-5
Molecular Formula C8H12Cl2N2O2
Molecular Weight (g/mol) 239.10
MDL Number MFCD13184932
InChI Key NAMMTAFKUFJMSD-UHFFFAOYNA-N
Synonym s-2-amino-3-pyridin-4-yl propanoic acid dihydrochloride, 3-4-pyridyl-l-alanine dihydrochloride, 2s-2-amino-3-pyridin-4-yl propanoic acid dihydrochloride, 3-pyridin-4-yl-l-alanine-hydrogen chloride 1/2, s-2-amino-3-pyridin-4-yl propanoic acid 2hcl, s-2-amino-3-pyridin-4-yl propanoicaciddihydrochloride
PubChem CID 20672030
SMILES Cl.Cl.NC(CC1=CC=NC=C1)C(O)=O

Specifications

IUPAC Name (2S)-2-amino-3-pyridin-4-ylpropanoic acid;dihydrochloride
Quantity 250 mg
Formula Weight 239.1
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-(4-Pyridyl)-L-alanine dihydrochloride
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.