Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4-Pyridyl)-L-alanine Dihydrochloride 96.0+%, TCI America™
SDP

Catalog No. P21361G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P21361G 1 g
P21365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P21361G Supplier TCI America Supplier No. P21361G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 178933-04-5
Molecular Formula C8H12Cl2N2O2
Molecular Weight (g/mol) 239.10
MDL Number MFCD13184932
InChI Key NAMMTAFKUFJMSD-UHFFFAOYNA-N
Synonym s-2-amino-3-pyridin-4-yl propanoic acid dihydrochloride, 3-4-pyridyl-l-alanine dihydrochloride, 2s-2-amino-3-pyridin-4-yl propanoic acid dihydrochloride, 3-pyridin-4-yl-l-alanine-hydrogen chloride 1/2, s-2-amino-3-pyridin-4-yl propanoic acid 2hcl, s-2-amino-3-pyridin-4-yl propanoicaciddihydrochloride
PubChem CID 20672030
IUPAC Name 2-amino-3-(pyridin-4-yl)propanoic acid dihydrochloride
SMILES Cl.Cl.NC(CC1=CC=NC=C1)C(O)=O

Specifications

Melting Point 206°C
Color White-Yellow
Quantity 1 g
Formula Weight 239.10
Percent Purity ≥96.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3-(4-Pyridyl)-L-alanine Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.