Learn More
Cayman Chemical 3- 4-PyrIdylIndole 10mg
SDP
Catalog No. 502968138
Encompass_Preferred
A ROCK1 inhibitor (IC50 = 25 µM) that promotes cell spreading, inhibits membrane blebbing, and induces dissolution of actin stress fibers in a wound healing assay; also inhibits ROCK2 and PRK2 with similar potency, while inhibiting MSK-1 and PKA with relatively weaker potency
Catalog No. 50-296-8138 Supplier Cayman Chemical Supplier No. 1792210 MG
A ROCK1 inhibitor (IC50 = 25 µM) that promotes cell spreading, inhibits membrane blebbing, and induces dissolution of actin stress fibers in a wound healing assay; also inhibits ROCK2 and PRK2 with similar potency, while inhibiting MSK-1 and PKA with relatively weaker potency