Learn More
Cayman Chemical 3-(4-Pyridyl)indole, Purity: ≥98%, CAS: 7272-84-6, 50 mg
SDP
Catalog No. 502965171
Encompass_Preferred
A ROCK1 inhibitor (IC50 = 25 µM) that promotes cell spreading, inhibits membrane blebbing, and induces dissolution of actin stress fibers in a wound healing assay; also inhibits ROCK2 and PRK2 with similar potency, while inhibiting MSK-1 and PKA with relatively weaker potency
Catalog No. 50-296-5171 Supplier Cayman Chemical Supplier No. 1792250 MG
A ROCK1 inhibitor (IC50 = 25 µM) that promotes cell spreading, inhibits membrane blebbing, and induces dissolution of actin stress fibers in a wound healing assay; also inhibits ROCK2 and PRK2 with similar potency, while inhibiting MSK-1 and PKA with relatively weaker potency