Learn More
COLDER PRODUCTS COMPANY AseptiQuik G Connector Body, High Temperature Hose Barb 3/4, USP Class VI ADCF, 5 per pack
Catalog No. 503131443
Encompass_Preferred
Colder Products Company AseptiQuik G Connector Body High Temperature Hose Barb 3/4 in White with white pull tab Flexibility Gamma autoclave Polycarbonate Silicone platinum-cured USP Class VI ADCF, 5 per pack
Catalog No. 50-313-1443 Supplier COLDER PRODUCTS COMPANY Supplier No. AQG33012HT
Colder Products Company AseptiQuik G Connector Body High Temperature Hose Barb 3/4 in White with white pull tab Flexibility Gamma autoclave Polycarbonate Silicone platinum-cured USP Class VI ADCF, 5 per pack