Learn More
COLDER PRODUCTS COMPANY Insert Coupling Colder Products Polysulfone USP Class VI ADCF 3/4 in hose barb Amber tint Free floating Up to 60 psi 4 1 bar -40 to 300 deg F (-40 tp 149 deg C) Gamma sterilized
Catalog No. 503131346
Encompass_Preferred
Insert Coupling Colder Products Polysulfone USP Class VI ADCF 3/4 in hose barb Amber tint Free floating Up to 60 psi 4 1 bar -40 to 300 deg F (-40 tp 149 deg C) Gamma sterilized
Catalog No. 50-313-1346 Supplier COLDER PRODUCTS COMPANY Supplier No. MPX44012T39M
Insert Coupling Colder Products Polysulfone USP Class VI ADCF 3/4 in hose barb Amber tint Free floating Up to 60 psi 4 1 bar -40 to 300 deg F (-40 tp 149 deg C) Gamma sterilized