Learn More
Medchemexpress LLC 3-(4-tert-butylphenyl)-2-methylpropanal | 80-54-6 | MFCD00047655 | 98.0% | 204.31 g/mol | C14H20O | 100 G
SDP
Catalog No. 5000302633
Encompass_Preferred
Lilial (3-(4-tert-butylphenyl)-2-methylpropanal, CAS 80-54-6) is an aromatic aldehyde used as a floral-fruity fragrance ingredient in perfumes, cosmetics, cleaning products, and air fresheners. It is a colorless to pale yellow liquid commonly supplied for formulation and laboratory use; typical purity is 98.0%.
  • Used as a floral-fruity fragrance ingredient in perfumes, cosmetics, and household products.
  • Contributes lily-like, powdery top notes at low concentrations.
  • High purity supports reproducible formulation and analytical testing.
  • Liquid form facilitates incorporation into liquid and emulsion systems.
  • Well-characterized chemical properties aid regulatory and safety assessment.
Catalog No. 50-003-02633 Supplier Medchemexpress LLC Supplier No. HYW014684100G
Lilial (3-(4-tert-butylphenyl)-2-methylpropanal, CAS 80-54-6) is an aromatic aldehyde used as a floral-fruity fragrance ingredient in perfumes, cosmetics, cleaning products, and air fresheners. It is a colorless to pale yellow liquid commonly supplied for formulation and laboratory use; typical purity is 98.0%.
  • Used as a floral-fruity fragrance ingredient in perfumes, cosmetics, and household products.
  • Contributes lily-like, powdery top notes at low concentrations.
  • High purity supports reproducible formulation and analytical testing.
  • Liquid form facilitates incorporation into liquid and emulsion systems.
  • Well-characterized chemical properties aid regulatory and safety assessment.