Learn More
Medchemexpress LLC 3',4'-Tetramethylene-α-pyrrolidinobutiophenone hydrochloride | 2749504-65-0 | 99.9% | 307.86 | 5 MG
SDP
Catalog No. 5000391026
Encompass_Preferred
This product is a laboratory chemical, 3',4'-Tetramethylene-α-pyrrolidinobutiophenone hydrochloride, intended for research use only and suitable for the manufacture of substances. It should be handled by suitably qualified and experienced scientists in appropriately equipped and authorized facilities. The substance is stable under recommended storage conditions but is incompatible with strong acids/alkalis and strong oxidizing/reducing agents. It is not classified as a hazardous substance or mixture.
  • Identified for laboratory use and substance manufacturing.
  • Stable under recommended storage conditions.
  • High purity of 99.9%.
  • Molecular formula C18H26CINO.
  • Powder storage: -20°C for 3 years, 4°C for 2 years.
  • In solvent storage: -80°C for 6 months, -20°C for 1 month.
  • Not a hazardous substance or mixture.
Catalog No. 50-003-91026 Supplier Medchemexpress LLC Supplier No. HY1684005MG
This product is a laboratory chemical, 3',4'-Tetramethylene-α-pyrrolidinobutiophenone hydrochloride, intended for research use only and suitable for the manufacture of substances. It should be handled by suitably qualified and experienced scientists in appropriately equipped and authorized facilities. The substance is stable under recommended storage conditions but is incompatible with strong acids/alkalis and strong oxidizing/reducing agents. It is not classified as a hazardous substance or mixture.
  • Identified for laboratory use and substance manufacturing.
  • Stable under recommended storage conditions.
  • High purity of 99.9%.
  • Molecular formula C18H26CINO.
  • Powder storage: -20°C for 3 years, 4°C for 2 years.
  • In solvent storage: -80°C for 6 months, -20°C for 1 month.
  • Not a hazardous substance or mixture.