Learn More
Chemglass Life Sciences 3/4" to 3/4" PFA Compression Fitting
SDP
Supplier:  Chemglass Life Sciences CG197523
Encompass_Preferred
3/4" to 3/4" PFA Compression Fitting. Designed for use with our CG-1975 heavy wall PTFE tubing. Fittings are made entirely of chemically inert corrosion resistant PFA PTFE. Sizes and configurations other than those listed are available.
Catalog No. CG197523