Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(4-(Tributylstannyl)thiazol-2-yl)oxetan-3-ol

Catalog No. SYX004371G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SYX004371G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SYX004371G Supplier Sigma Aldrich Supplier No. SYX004371G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-fluoroanisole was used in the synthesis of 4-fluoro-5,6-dihydroxytryptamine
  • It was also used in the synthesis of 3-fluoro- and 5-fluoronoradrenaline

Specifications

CAS 1245816-13-0
Molecular Formula C18H33NO2SSn
Linear Formula C18H33NO2SSn
MDL Number MFCD16140389
Quantity 25 g
Molecular Weight (g/mol) 446.24
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.