Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-[4-(Trifluoromethoxy)phenyl]acrylic acid, 97%, Thermo Scientific™

Catalog No. SEW04310DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SEW04310DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SEW04310DA Supplier Thermo Scientific Chemicals Supplier No. SEW04310DA
Add to Cart

Chemical Identifiers

CAS 783-13-1
Molecular Formula C10H7F3O3
Molecular Weight (g/mol) 232.158
MDL Number MFCD00066338
InChI Key RNYVTJANWYBGPW-ZZXKWVIFSA-N
Synonym 3-4-trifluoromethoxy phenyl acrylic acid, 4-trifluoromethoxy cinnamic acid, e-3-4-trifluoromethoxy phenyl acrylic acid, e-3-4-trifluoromethoxy phenyl prop-2-enoic acid, 2e-3-4-trifluoromethoxy phenyl prop-2-enoic acid, 3-4-trifluoromethoxyphenyl acrylic acid, trifluoromethoxyphenylacrylicacid, trans-4-trifluoromethoxy cinnamic acid
PubChem CID 735857
IUPAC Name (E)-3-[4-(trifluoromethoxy)phenyl]prop-2-enoic acid
SMILES C1=CC(=CC=C1C=CC(=O)O)OC(F)(F)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 232.16
Percent Purity 97%
Chemical Name or Material 3-[4-(Trifluoromethoxy)phenyl]acrylic acid
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.