Learn More
Holland Applied Technologies 3/4 USP CL VI EPDM SANITARY GS
Catalog No. NC9435568
Encompass_Preferred
This item has a minimum order quantity of 100 pieces per supplier requirements
0.75 in. USP CLASS VI EPDM GASKET, LOT NUMBER LASER ETCHED ON SIDE
Catalog No. NC9435568 Supplier Holland Applied Technologies Supplier No. 42MPE075LE
This item has a minimum order quantity of 100 pieces per supplier requirements
0.75 in. USP CLASS VI EPDM GASKET, LOT NUMBER LASER ETCHED ON SIDE