Learn More
MSC Dymo Vinyl Tape, 3/4 in, White, 18 ft, 1 ea
Catalog No. 501113156
Encompass
DYMO Industrial Vinyl tape features a permanent acrylic adhesive and topcoat for thermal transfer printing. Designed for general ID, electrical panels, asset labeling, and pipe marking, it is suitable for both indoor and outdoor use on curved or flat surfaces. This halogen-free tape offers resistance to moisture, chemicals, and UV.
  • Color-coded for safety standards (OSHA, ANSI, ISO)
  • Flexible for curved, textured, or flat surfaces
  • Scratch, tear, and abrasion-resistant text
  • Withstands moisture, UV light, oil, solvents, and temperatures from -40°C to +80°C
  • UL recognized and meets REACH/ROHS standards
Catalog No. 50-111-3156 Supplier MSC Supplier No. 93427276
DYMO Industrial Vinyl tape features a permanent acrylic adhesive and topcoat for thermal transfer printing. Designed for general ID, electrical panels, asset labeling, and pipe marking, it is suitable for both indoor and outdoor use on curved or flat surfaces. This halogen-free tape offers resistance to moisture, chemicals, and UV.
  • Color-coded for safety standards (OSHA, ANSI, ISO)
  • Flexible for curved, textured, or flat surfaces
  • Scratch, tear, and abrasion-resistant text
  • Withstands moisture, UV light, oil, solvents, and temperatures from -40°C to +80°C
  • UL recognized and meets REACH/ROHS standards