Learn More
Cayman Chemical 3 4DHMA trifluoroacetate sal
SDP
Supplier:  Cayman Chemical 105155 MG
Encompass_Preferred
An analytical reference standard categorized as an amphetamine; an active metabolite of 3,4-MDMA; intended for research and forensic applications
Catalog No. 50-288-9662