Learn More
Cayman Chemical 3,4-Dihydroxymandelic Acid, Purity: ≥95%, CAS: 775-01-9, 250 mg
SDP
Catalog No. 502912221
Encompass_Preferred
An active metabolite of norepinephrine; scavenges superoxide radicals (IC50 = 130 nM) and reduces lipid peroxidation in cell-free assays; a chemoattractant for enterohemorrhagic E. coli; induces the expression of enterohemorrhagic E. coli virulence genes at 50 µM; increases the attachment of enterohemorrhagic E. coli to HeLa cells in vitro
Catalog No. 50-291-2221 Supplier Cayman Chemical Supplier No. 36369250 MG
An active metabolite of norepinephrine; scavenges superoxide radicals (IC50 = 130 nM) and reduces lipid peroxidation in cell-free assays; a chemoattractant for enterohemorrhagic E. coli; induces the expression of enterohemorrhagic E. coli virulence genes at 50 µM; increases the attachment of enterohemorrhagic E. coli to HeLa cells in vitro