Learn More
Cayman Chemical 3,4-Dihydroxyphenyl ethanol, Purity: ≥98%, CAS: 10597-60-1, 50 mg
SDP
Catalog No. 502895274
Encompass_Preferred
3,4-Dihydroxyphenyl ethanol is a phenolic component of olive oil that inhibits both 12- and 5-LO. The IC50 values for the inhibition of rat platelet 12-LO and rat neutrophil 5-LO are 4.2 and 13 µM, respectively. It does not inhibit, and may actually enhance, COX activity.{4049} 3,4-Dihydroxyphenyl ethanol also protects LDL from both biological and chemical oxidation, suggesting a potential mechanism for the protective effects of olive oil against atherosclerosis.{6597}
Catalog No. 50-289-5274 Supplier Cayman Chemical Supplier No. 7060450 MG
3,4-Dihydroxyphenyl ethanol is a phenolic component of olive oil that inhibits both 12- and 5-LO. The IC50 values for the inhibition of rat platelet 12-LO and rat neutrophil 5-LO are 4.2 and 13 µM, respectively. It does not inhibit, and may actually enhance, COX activity.{4049} 3,4-Dihydroxyphenyl ethanol also protects LDL from both biological and chemical oxidation, suggesting a potential mechanism for the protective effects of olive oil against atherosclerosis.{6597}