Learn More
Cayman Chemical 3,4-dimethoxy-α-Pyrrolidinopentiophenone (hydrochloride), Purity: ≥98%, CAS: 850351-99-4, 50 mg
SDP
Catalog No. 502953409
Encompass_Preferred
An analog of MDPV, differing only by an opening of the ring structure on the phenyl group; intended for research and forensic applications
Catalog No. 50-295-3409 Supplier Cayman Chemical Supplier No. 1410650 MG
An analog of MDPV, differing only by an opening of the ring structure on the phenyl group; intended for research and forensic applications