Learn More
Hach Company 3/4in Comb pH Sen-Convert Flat
Catalog No. 5000207311 Shop All Hach Company Products
Encompass
3/4 Inch Combination pH/ORP Sensor-Convertible Sensor Style-PPS Body Material-Flat Glass General Purpose pH ElectrodePPS Convertible Body Style 4 5m Analog Cable General Purpose Flat Glass pH Electrode No Temperature Compensation
Catalog No. 50-002-07311 Supplier Hach Company Supplier No. PC1R2N
3/4 Inch Combination pH/ORP Sensor-Convertible Sensor Style-PPS Body Material-Flat Glass General Purpose pH ElectrodePPS Convertible Body Style 4 5m Analog Cable General Purpose Flat Glass pH Electrode No Temperature Compensation