Learn More
Hach Company 3/4in Comb pH Sen-Convert Gls
Catalog No. 5000207593 Shop All Hach Company Products
Encompass
3/4in Combination pH Sensor-Convertible Style-PPS-General Purpose Glass pH Electrode Type-Temperature CompensationPPS Body Material convertible body style 4 5 m analog cable general purpose glass pH electrode PT100 temperature compensation
Catalog No. 50-002-07593 Supplier Hach Company Supplier No. PC1R1AV12
3/4in Combination pH Sensor-Convertible Style-PPS-General Purpose Glass pH Electrode Type-Temperature CompensationPPS Body Material convertible body style 4 5 m analog cable general purpose glass pH electrode PT100 temperature compensation