Learn More
Hach Company 3/4in Comb pH Sen-Incert Flat
Catalog No. 5000207410 Shop All Hach Company Products
Encompass
3/4 Inch Combination pH / ORP Sensor-Insertion Style-PVDF Body Material-Flat Glass pH ElectrodePVDF Insertion Body Style 4 5m Analog Cable Flat Glass pH Electrode Pt 1000 Temperature Compensation
Catalog No. 50-002-07410 Supplier Hach Company Supplier No. PC2K2A
3/4 Inch Combination pH / ORP Sensor-Insertion Style-PVDF Body Material-Flat Glass pH ElectrodePVDF Insertion Body Style 4 5m Analog Cable Flat Glass pH Electrode Pt 1000 Temperature Compensation