Learn More
Hach Company 3/4in Comb pH Sen-Incert Glass
Catalog No. 5000207499 Shop All Hach Company Products
Encompass
3/4 Inch Combination pH/ORP Sensor-Insertion Style-PVDF Body Material-General Purpose Glass pH ElectrodePVDF Insertion Body Style 4 5m Analog Cable General Purpose Glass pH Electrode Pt 1000 Temperature Compensation
Catalog No. 50-002-07499 Supplier Hach Company Supplier No. PC2K1A
3/4 Inch Combination pH/ORP Sensor-Insertion Style-PVDF Body Material-General Purpose Glass pH ElectrodePVDF Insertion Body Style 4 5m Analog Cable General Purpose Glass pH Electrode Pt 1000 Temperature Compensation