Learn More
Hach Company 3/4in Comb pH Sen-Sani SS/PVDF
Catalog No. 5000207411 Shop All Hach Company Products
Encompass
3/4 Inch Combination pH/ORP Sensor-Sanitary Sensor Style-316 SS/PVDF Body Material-General Purpose pH Glass Electrode316 SSVDF Sanitary Body Style 4 5m Analog Cable General Purpose Glass pH Electrode Pt 1000 Temperature Compensation
Catalog No. 50-002-07411 Supplier Hach Company Supplier No. PC3K2A
3/4 Inch Combination pH/ORP Sensor-Sanitary Sensor Style-316 SS/PVDF Body Material-General Purpose pH Glass Electrode316 SSVDF Sanitary Body Style 4 5m Analog Cable General Purpose Glass pH Electrode Pt 1000 Temperature Compensation