Learn More
MSC Value Collection Machine Screw, 8-32 x 3/4 in, 1 ea
Catalog No. NC1895610
Encompass
This machine screw, part of the Value Collection, features an #8-32 thread and a 3/4 inch length under the head. Constructed from 18-8 grade stainless steel, it is designed for secure fastening in various applications.
  • Thread size: #8-32
  • Length under head: 3/4 in
  • Head type: Pan
  • Drive type: Phillips
  • Material: 18-8 grade stainless steel
  • Finish: Bright/uncoated
  • Threading: UNC
  • Washer style: Without washer
Catalog No. NC1895610 Supplier MSC Supplier No. 51251197
This machine screw, part of the Value Collection, features an #8-32 thread and a 3/4 inch length under the head. Constructed from 18-8 grade stainless steel, it is designed for secure fastening in various applications.
  • Thread size: #8-32
  • Length under head: 3/4 in
  • Head type: Pan
  • Drive type: Phillips
  • Material: 18-8 grade stainless steel
  • Finish: Bright/uncoated
  • Threading: UNC
  • Washer style: Without washer