Learn More
Cayman Chemical 3,4-Methylenedioxy-5-methylethcathinone (hydrochloride), Purity: ≥95%, CAS: 2748289-02-1, 10 mg
SDP
Catalog No. 502952829
Encompass_Preferred
An analog of the cathinone bk-MDEA, differing by the addition of a methyl group at the two position of the benzene ring
Catalog No. 50-295-2829 Supplier Cayman Chemical Supplier No. 1407810 MG
An analog of the cathinone bk-MDEA, differing by the addition of a methyl group at the two position of the benzene ring